Anti-CXCL14 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-811
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CXCL14
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:O95715
- Antigen Synonyms:1110031L23Rik|1200006I23Rik|AI414372|BMAC|bolekine|BRAK|Breast and kidney|C-X-C motif chemokine 14|C-X-C motif chemokine ligand 14|Chaemokine|CXC motif|ligand 14|Chemokine (C-X-C motif) ligand 14|Chemokine BRAK|CXC chemokine in breast and kidney |CXCL14|CXL14_HUMAN|JSC|Kec|Kidney-expressed chemokine CXC|KS1|MGC10687|MGC124510|MGC90667|MIP 2 gamma|MIP-2G|MIP2G|MIP2gamma|NJAC|PRO273|PSEC0212|Scyb14|Small Inducible Cytokine B14|Small inducible cytokine subfamily B (Cys-X-Cys) member 14 (BRAK)|Small Inducible Cytokine subfamily B|member 14|Small-inducible cytokine B14|Tumor suppressing chemokine|UNQ240
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human C-X-C motif chemokine 14 protein (35-111AA)
- Purification:Antigen affinity purified
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human