Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Factor H
Catalog # 80051-582
Supplier:  MilliporeSigma
CAS Number:  
Human Factor H
Catalog # 80051-582
Supplier:  MilliporeSigma
Supplier Number:  341274-250UG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic ampoule
  • Species:
    Human
  • Size:
    250 μg
  • Protein Synonyms:
    β 1H Globulin|β1H Globulin
  • Protein/Peptide Name:
    Factor H
  • Purity:
    ≥95% (SDS-PAGE)

Specifications

About this item

Factor H, Human, Factor H, Human, is a single-chain multi-functional glycoprotein that regulates formation and function of complement C3 and C5 convertase enzymes.

Native factor H from human serum. Single-chain glycoprotein present in normal human serum at about 500 µg/ml. Regulates formation and function of complement C3 and C5 convertase enzymes. A C3b-binding protein, not an enzyme. Regulatory activity is attributed to its ability to recognize and bind to C3b fragments.