Anti-NDEL1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: NDEL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77849-061
- 77849-060
- Antigen Symbol:
- NDEL1
- NDEL1
- Antigen Synonyms:
- A. nidulans|DKFZp451M0318|ENDOOLIGOPEPTIDASE A|EOPA|MITAP 1|MITAP1|Mitosin associated protein 1|Mitosin associated protein MITAP1|Mitosin-associated protein 1|Ndel 1|NDEL1_HUMAN|Nuclear distribution gene E like homolog 1|Nuclear distribution protein nudE like 1|Nuclear distribution protein nudE-like 1|NUDE like protein|NudE nuclear distribution gene E homolog like 1 A. nidulans|NudE nuclear distribution gene E homolog like 1|NUDEL|Protein Nudel
- A. nidulans|DKFZp451M0318|ENDOOLIGOPEPTIDASE A|EOPA|MITAP 1|MITAP1|Mitosin associated protein 1|Mitosin associated protein MITAP1|Mitosin-associated protein 1|Ndel 1|NDEL1_HUMAN|Nuclear distribution gene E like homolog 1|Nuclear distribution protein nudE like 1|Nuclear distribution protein nudE-like 1|NUDE like protein|NudE nuclear distribution gene E homolog like 1 A. nidulans|NudE nuclear distribution gene E homolog like 1|NUDEL|Protein Nudel
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9GZM8
- Q9GZM8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human NDEL1
- Fusion protein of Human NDEL1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice