Anti-UBE2D4 Rabbit Recombinant Polyclonal Antibody
New Product
Type:
Antigen: UBE2D4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77845-397
- 77845-398
- Antigen Symbol:
- UBE2D4
- UBE2D4
- Antigen Synonyms:
- UBE2D4|UBCH5D|Ubiquitin-conjugating enzyme E2 D4|EC 2.3.2.23|E2 ubiquitin-conjugating enzyme D4|HBUCE1|Ubiquitin carrier protein D4|Ubiquitin-protein ligase D4
- UBE2D4|UBCH5D|Ubiquitin-conjugating enzyme E2 D4|EC 2.3.2.23|E2 ubiquitin-conjugating enzyme D4|HBUCE1|Ubiquitin carrier protein D4|Ubiquitin-protein ligase D4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2X8
- Q9Y2X8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ubiquitin-conjugating enzyme E2 D4 protein (1-147AA)
- Recombinant Human Ubiquitin-conjugating enzyme E2 D4 protein (1-147AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice