Anti-RAPGEF2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RAPGEF2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-386
- 77846-387
- Antigen Symbol:
- RAPGEF2
- RAPGEF2
- Antigen Synonyms:
- CNrasGEF|Cyclic nucleotide ras GEF|KIAA0313|Neural RAP guanine nucleotide exchange protein|nRap GEP|NRAPGEP|PDZ domain containing guanine nucleotide exchange factor 1|PDZ domain-containing guanine nucleotide exchange factor 1|PDZ GEF1|PDZ-GEF1|PDZGEF1|RA GEF|RA-GEF-1|Rap guanine nucleotide exchange factor 2|Rapgef2|Ras/Rap1-associating GEF-1|RPGF2_HUMAN
- CNrasGEF|Cyclic nucleotide ras GEF|KIAA0313|Neural RAP guanine nucleotide exchange protein|nRap GEP|NRAPGEP|PDZ domain containing guanine nucleotide exchange factor 1|PDZ domain-containing guanine nucleotide exchange factor 1|PDZ GEF1|PDZ-GEF1|PDZGEF1|RA GEF|RA-GEF-1|Rap guanine nucleotide exchange factor 2|Rapgef2|Ras/Rap1-associating GEF-1|RPGF2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y4G8
- Q9Y4G8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Rap guanine nucleotide exchange factor 2 protein (1393-1498AA)
- Recombinant Human Rap guanine nucleotide exchange factor 2 protein (1393-1498AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice