Anti-HEBP2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: HEBP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-476
- 77845-477
- Antigen Symbol:
- HEBP2
- HEBP2
- Antigen Synonyms:
- C6orf34|Chromosome 6 open reading frame 34|HEBP2_HUMAN|Heme binding protein 2|Heme-binding protein 2|Placental protein 23|PP23|Protein SOUL|SOUL
- C6orf34|Chromosome 6 open reading frame 34|HEBP2_HUMAN|Heme binding protein 2|Heme-binding protein 2|Placental protein 23|PP23|Protein SOUL|SOUL
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5Z4
- Q9Y5Z4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Heme-binding protein 2 protein (2-205AA)
- Recombinant Human Heme-binding protein 2 protein (2-205AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice