Anti-CLCA1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-180
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CLCA1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:A8K7I4
- Antigen Synonyms:CaCC| CaCC-1| CACC1| Calcium dependent chloride channel 1| Calcium-activated chloride channel family member 1| Calcium-activated chloride channel protein 1| Calcium-activated chloride channel regulator 1| Chloride channel calcium activated family member 1| Chloride channel regulator 1| CLCA family member 1 chloride channel regulator| CLCA1| CLCA1_HUMAN| CLCRG1| GOB5| hCaCC-1| hCLCA1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Calcium-activated chloride channel regulator 1 protein (715-785AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: CLCA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human