Anti-AP2B1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-222
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AP2B1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P63010
- Antigen Synonyms:Adapter related protein complex 2 beta 1 subunit|Adapter-related protein complex 2 beta subunit|Adaptin|beta 2 (beta)|Adaptor protein complex AP-2 subunit beta|Adaptor related protein complex 2|beta 1 subunit|ADTB2|AP-2 complex subunit beta|AP105B|AP2 BETA|AP2B1_HUMAN|Beta adaptin|Beta-2-adaptin|Beta-adaptin|Beta2 adaptin|CLAPB1|Clathrin assembly protein complex 2 beta large chain|Clathrin associated/assembly/adaptor protein|large|beta 1|DKFZp781K0743|Plasma membrane adaptor HA2/AP2 adaptin beta subunit
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human AP-2 complex subunit beta protein (738-937AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: AP2B1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse