Anti-HEY2 Rabbit Polyclonal Antibody
77849-902
New Product
- Antigen Symbol:HEY2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:liquid
- Gene ID:Q9UBP5
- Antigen Synonyms:bHLHb32|Cardiovascular helix loop helix factor 1|Cardiovascular helix-loop-helix factor 1|CHF1|Class B basic helix-loop-helix protein 32|GRIDLOCK|GRL|Hairy and enhancer of split related 2|Hairy and enhancer of split-related protein 2|Hairy related transcription factor 2|Hairy-related transcription factor 2|Hairy/enhancer-of-split related with YRPW motif protein 2|hCHF1|HERP|HERP1|HES related repressor protein 1|HES-related repressor protein 2|HESR-2|HESR2|HEY2_HUMAN|hHRT2|HRT-2|HRT2|MGC10720|Protein gridlock homolog
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human HEY2.
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: HEY2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse