Anti-DHRS9 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
77854-170
New Product
- Antigen Symbol:DHRS9
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9BPW9
- Antigen Synonyms:3 alpha hydroxysteroid dehydrogenase| 3-@alpha-HSD| 3-@alpha-hydroxysteroid dehydrogenase| 3-alpha hydroxysteroid dehydrogenase| 3-alpha-HSD| 3alpha HSD| Dehydrogenase/reductase SDR family member 9| DHRS9| DHRS9_HUMAN| NADP dependent retinol dehydrogenase/reductase| NADP-dependent retinol dehydrogenase/reductase| RDH-E2| Rdh15| RDHE2| RDHL| RDHTBE| retinol dehydrogenase homolog | Retinol dehydrogenase| tracheobronchiol epithelail cell-specific| RETSDR8| SDR family| member 9| SDR9C4| short chain dehydrogenase/reductase family 9| short chain dehydrogenase/reductase family 9C| member 4 | Short chain dehydrogenase/reductase retSDR8| Short-chain dehydrogenase/reductase retSDR8
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Dehydrogenase/reductase SDR family member 9 protein (23-319AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: DHRS9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human