Anti-TENM1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TENM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-925
- 77844-926
- Antigen Symbol:
- TENM1
- TENM1
- Antigen Synonyms:
- ODZ1|TNM1|Teneurin-1|Ten-1|Protein Odd Oz/ten-m homolog 1|Tenascin-M1|Ten-m1|Teneurin transmembrane protein 1) [Cleaved into: Ten-1 intracellular domain|IDten-1|Ten-1 ICD)|Teneurin C-terminal-associated peptide|TCPA-1|Ten-1 extracellular domain|Ten-1 ECD)]
- ODZ1|TNM1|Teneurin-1|Ten-1|Protein Odd Oz/ten-m homolog 1|Tenascin-M1|Ten-m1|Teneurin transmembrane protein 1) [Cleaved into: Ten-1 intracellular domain|IDten-1|Ten-1 ICD)|Teneurin C-terminal-associated peptide|TCPA-1|Ten-1 extracellular domain|Ten-1 ECD)]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UKZ4
- Q9UKZ4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Teneurin-1 protein (1-241AA)
- Recombinant human Teneurin-1 protein (1-241AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice