Anti-RNF6 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: RNF6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-056
- 77846-057
- Antigen Symbol:
- RNF6
- RNF6
- Antigen Synonyms:
- E3 ubiquitin-protein ligase Ring finger protein (C3H2C3 type) 6|Ring finger protein 6|RING H2 protein|RING H2 protein RNF 6|RNF6_HUMAN|SPG2
- E3 ubiquitin-protein ligase Ring finger protein (C3H2C3 type) 6|Ring finger protein 6|RING H2 protein|RING H2 protein RNF 6|RNF6_HUMAN|SPG2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y252
- Q9Y252
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human E3 ubiquitin-protein ligase RNF6 protein (1-298AA)
- Recombinant Human E3 ubiquitin-protein ligase RNF6 protein (1-298AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice