Anti-ACER1 Rabbit Polyclonal Antibody
Catalog # 77846-969
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ACER1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q8TDN7
- Antigen Synonyms:ACER1_HUMAN|Acylsphingosine deacylase 3|Alkaline CDase 1|Alkaline ceramidase 1|Alkaline ceramidase|AlkCDase 1|ALKCDase1|ASAH 3|MGC138327|MGC138329|N acylsphingosine amidohydrolase (alkaline ceramidase) 3|N acylsphingosine amidohydrolase 3 alkaline ceramidase|N acylsphingosine amidohydrolase 3|N-acylsphingosine amidohydrolase 3
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human ASAH3. at AA rangle: 100-180
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:2000, ELISA:1:10000
Type:
Antigen: ACER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human