Anti-CDC42EP2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77852-810
New Product
- Antigen Symbol:CDC42EP2
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O14613
- Antigen Synonyms:Binder of Rho GTPases 1|BORG1|BORG1_HUMAN|CDC42 effector protein (Rho GTPase binding) 2|Cdc42 effector protein 2|CEP2|CRIB-containing BOGR1 protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cdc42 effector protein 2 protein (9-210AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: CDC42EP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human