Anti-VAV3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: VAV3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-757
- 77844-758
- Antigen Symbol:
- VAV3
- VAV3
- Antigen Synonyms:
- FLJ40431|Guanine nucleotide exchange factor Protein vav 3|Protein RGD1565941|VAV 3|Vav 3 guanine nucleotide exchange factor|VAV 3 oncogene|VAV 3 protein|VAV-3|VAV3 oncogene|VAV3 protein|VAV3_HUMAN
- FLJ40431|Guanine nucleotide exchange factor Protein vav 3|Protein RGD1565941|VAV 3|Vav 3 guanine nucleotide exchange factor|VAV 3 oncogene|VAV 3 protein|VAV-3|VAV3 oncogene|VAV3 protein|VAV3_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UKW4
- Q9UKW4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Guanine nucleotide exchange factor VAV3 protein (562-674AA)
- Recombinant Human Guanine nucleotide exchange factor VAV3 protein (562-674AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together