Anti-ADM Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: ADM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-706
- 77843-705
- Antigen Symbol:
- ADM
- ADM
- Antigen Synonyms:
- ADM precursor|ADML_HUMAN|Adrenomedullin|AM|PAMP|Preproadrenomedullin|Proadrenomedullin N-20 terminal peptide|Proadrenomedullin N20 terminal peptide|ProAM N terminal 20 peptide|ProAM N-terminal 20 peptide|ProAM N20|ProAM-N20|ProAMN20
- ADM precursor|ADML_HUMAN|Adrenomedullin|AM|PAMP|Preproadrenomedullin|Proadrenomedullin N-20 terminal peptide|Proadrenomedullin N20 terminal peptide|ProAM N terminal 20 peptide|ProAM N-terminal 20 peptide|ProAM N20|ProAM-N20|ProAMN20
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P35318
- P35318
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human ADM
- Synthetic peptide of Human ADM
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice