Anti-NRAS Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: NRAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77849-535
- 77849-534
- Antigen Symbol:
- NRAS
- NRAS
- Antigen Synonyms:
- ALPS4|AV095280|GTPase NRas|HRAS1|N ras|N ras protein part 4|Neuroblastoma RAS viral (v ras) oncogene homolog|NRAS|NRAS1|NS6|OTTHUMP00000013879|OTTMUSP00000023521|RASN_HUMAN|Transforming protein N Ras|Transforming protein N-Ras|v ras neuroblastoma RAS viral oncogene homolog
- ALPS4|AV095280|GTPase NRas|HRAS1|N ras|N ras protein part 4|Neuroblastoma RAS viral (v ras) oncogene homolog|NRAS|NRAS1|NS6|OTTHUMP00000013879|OTTMUSP00000023521|RASN_HUMAN|Transforming protein N Ras|Transforming protein N-Ras|v ras neuroblastoma RAS viral oncogene homolog
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P01111
- P01111
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human NRAS
- Synthetic peptide of Human NRAS
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice