Anti-ADRA1A Rabbit Polyclonal Antibody (Biotinylated)
Catalog # 77850-823
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ADRA1A
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P35348
- Antigen Synonyms:ADA1A_HUMAN|Adra1c|ADRA1L1|Adrenergic alpha1A receptor|Adrenergic alpha1C receptor|Adrenergic receptor alpha 1a|Adrenergic receptor alpha 1c|Adrenergic|alpha 1A|receptor variant 1|receptor variant 11|receptor variant 3|receptor variant 5|receptor variant 8|Adrenoceptor alpha 1A|Alpha 1A adrenoreceptor|Alpha 1C adrenergic receptor|Alpha adrenergic receptor 1c|Alpha-1A adrenergic receptor|Alpha-1A adrenoceptor|Alpha-1A adrenoreceptor|Alpha-1C adrenergic receptor|Alpha-adrenergic receptor 1c|Alpha1A adrenergic receptor|ALPHA1AAR|G protein coupled receptor
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Peptide sequence from Human Alpha-1A adrenergic receptor protein (6-22AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human