Anti-ACVR1B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77850-708
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ACVR1B
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P36896
- Antigen Synonyms:Activin A receptor type 1B|Activin A receptor type II like kinase 4|Activin A type 1B receptor|Activin A type IB receptor|Activin receptor like kinase 4|Activin receptor type 1B|Activin receptor type IB|Activin receptor type-1B|Activin receptor-like kinase 4|ACTR IB|ACTR-IB|ACTRIB|ACV1B_HUMAN|ACVR 1B|ACVRLK 4|ACVRLK4|ALK 4|ALK-4|ALK4|Serine(threonine) protein kinase receptor R2|Serine/threonine-protein kinase receptor R2|SKR 2|SKR2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Activin receptor type-1B protein (150-300AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ACVR1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human