Anti-AS3MT Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: AS3MT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-266
- 77843-265
- Antigen Symbol:
- AS3MT
- AS3MT
- Antigen Synonyms:
- 2310045H08Rik|Arsenic (+3 oxidation state) methyltransferase|Arsenite methyltransferase|As3mt|AS3MT_HUMAN|C10ORF32|CYT19|Cyt19 protein|Hypothetical protein C10orf32|Methylarsonite methyltransferase|Methyltransferase cyt19|OTTHUMP00000020384|RP11-753C18.6|S adenosylmethionine arsenic (III) methyltransferase|S-adenosyl-L-methionine:arsenic(III) methyltransferase
- 2310045H08Rik|Arsenic (+3 oxidation state) methyltransferase|Arsenite methyltransferase|As3mt|AS3MT_HUMAN|C10ORF32|CYT19|Cyt19 protein|Hypothetical protein C10orf32|Methylarsonite methyltransferase|Methyltransferase cyt19|OTTHUMP00000020384|RP11-753C18.6|S adenosylmethionine arsenic (III) methyltransferase|S-adenosyl-L-methionine:arsenic(III) methyltransferase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HBK9
- Q9HBK9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Arsenite methyltransferase protein (262-375AA)
- Recombinant Human Arsenite methyltransferase protein (262-375AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice