Anti-ACOT8 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77850-650
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ACOT8
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:O14734
- Antigen Synonyms:ACTEIII|PTE1|Acyl-coenzyme A thioesterase 8|Acyl-CoA thioesterase 8|EC 3.1.2.1|EC 3.1.2.11|EC 3.1.2.2|EC 3.1.2.3|EC 3.1.2.5|Choloyl-coenzyme A thioesterase|EC 3.1.2.27|HIV-Nef-associated acyl-CoA thioesterase|Peroxisomal acyl-CoA thioesterase 2|PTE-2|Peroxisomal acyl-coenzyme A thioester hydrolase 1|PTE-1|Peroxisomal long-chain acyl-CoA thioesterase 1|Thioesterase II|hACTE-III|hACTEIII|hTE
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Acyl-coenzyme A thioesterase 8 protein (1-200AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ACOT8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human