Anti-PIGL Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PIGL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-291
- 77846-290
- Antigen Symbol:
- PIGL
- PIGL
- Antigen Synonyms:
- N acetylglucosaminylphosphatidylinositol deacetylase|N-acetylglucosaminyl-phosphatidylinositol de-N-acetylase|Phosphatidylinositol glycan anchor biosynthesis class L|Phosphatidylinositol glycan class L|Phosphatidylinositol-glycan biosynthesis class L protein|PIG-L|Pigl|PIGL_HUMAN
- N acetylglucosaminylphosphatidylinositol deacetylase|N-acetylglucosaminyl-phosphatidylinositol de-N-acetylase|Phosphatidylinositol glycan anchor biosynthesis class L|Phosphatidylinositol glycan class L|Phosphatidylinositol-glycan biosynthesis class L protein|PIG-L|Pigl|PIGL_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human N-acetylglucosaminyl-phosphatidylinositol de-N-acetylase protein (23-252AA)
- Recombinant Human N-acetylglucosaminyl-phosphatidylinositol de-N-acetylase protein (23-252AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice