Anti-SLC4A1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:15 to 1:50
Type:
Antigen: SLC4A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77848-664
- 77848-663
- Antigen Symbol:
- SLC4A1
- SLC4A1
- Antigen Synonyms:
- AE 1| AE1| Anion exchange protein 1| Anion exchanger 1| B3AT_HUMAN| Band 3 anion transport protein| Band 3| BND3| CD233| DI| Diego blood group| EMPB3| EPB3| Erythrocyte membrane protein band 3| Erythroid anion exchange protein| FR| Froese blood group| RTA1A| Solute carrier family 4 anion exchanger member 1| Solute carrier family 4 member 1| SW| Swann blood group| Waldner blood group| WD| WD1| WR| Wright blood group
- AE 1| AE1| Anion exchange protein 1| Anion exchanger 1| B3AT_HUMAN| Band 3 anion transport protein| Band 3| BND3| CD233| DI| Diego blood group| EMPB3| EPB3| Erythrocyte membrane protein band 3| Erythroid anion exchange protein| FR| Froese blood group| RTA1A| Solute carrier family 4 anion exchanger member 1| Solute carrier family 4 member 1| SW| Swann blood group| Waldner blood group| WD| WD1| WR| Wright blood group
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P02730
- P02730
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SLC4A1
- Synthetic peptide of Human SLC4A1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice