Anti-IFNA21 Rabbit Recombinant Polyclonal Antibody
Catalog # 77857-882
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:IFNA21
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P01568
- Antigen Synonyms:Interferon alpha-21|IFN-alpha-21|Interferon alpha-F|LeIF F
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Interferon alpha-21 protein (24-189AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IP: 1:200 to 1:2000
Type:
Antigen: IFNA21
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse