Anti-CORO1C Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CORO1C
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-652
- 77844-653
- Antigen Symbol:
- CORO1C
- CORO1C
- Antigen Synonyms:
- COR1C_HUMAN|CORO 1C|Coronin 1c|Coronin actin binding protein 1C|Coronin-1C|Coronin-3|Coronin1c|Coronin3|CRNN 4|CRNN4|HCRNN 4|hCRNN4
- COR1C_HUMAN|CORO 1C|Coronin 1c|Coronin actin binding protein 1C|Coronin-1C|Coronin-3|Coronin1c|Coronin3|CRNN 4|CRNN4|HCRNN 4|hCRNN4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULV4
- Q9ULV4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Coronin-1C protein (351-464AA)
- Recombinant Human Coronin-1C protein (351-464AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice