Anti-mib1 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: mib1
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
- Catalog No:
- 77845-299
- 77845-298
- Antigen Symbol:
- mib1
- mib1
- Antigen Synonyms:
- E3 ubiquitin-protein ligase mib1 (EC 2.3.2.27) (Protein mind bomb) (RING-type E3 ubiquitin transferase mib1), mib1, mib
- E3 ubiquitin-protein ligase mib1 (EC 2.3.2.27) (Protein mind bomb) (RING-type E3 ubiquitin transferase mib1), mib1, mib
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Zebrafish
- Zebrafish
- Host:
- Rabbit
- Rabbit
- Gene ID:
- H9GYU5
- H9GYU5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Zebrafish E3 ubiquitin-protein ligase mib1 protein (1-274AA)
- Recombinant Zebrafish E3 ubiquitin-protein ligase mib1 protein (1-274AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice