Streptomyces sp. N174 Chitosanase
80051-012
:
- Pack type:Plastic ampoule
- CAS No.:51570-20-8
- Species:Streptomyces sp. N174
- Size:10 U
- Enzyme Name:Chitosanase
- Enzyme Synonyms:Chitosanase; Chitosan N-acetylglucosaminohydrolase
Chitosanase, Streptomyces sp. N174, Chitosanase, Streptomyces sp. N174, CAS 51570-20-8, is a member of a new class of enzymes with antifungal properties. Efficiently catalyzes the hydrolysis of acetylated chitosan (0 to 60%).
Chitosanase, Streptomyces sp. N174, CAS 51570-20-8, is a member of a new class of enzymes with antifungal properties. Efficiently catalyzes the hydrolysis of acetylated chitosan (0 to 60%).