Anti-ALAS2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77850-998
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ALAS2
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P22557
- Antigen Synonyms:5 @aminolevulinate synthase erythroid specific|5 aminolevulinate synthase 2|5 aminolevulinate synthase 5 aminolevulinate synthase 2|5 aminolevulinate synthase erythroid specific mitochondrial|5 aminolevulinic acid synthase 2|5 aminolevulinic acid synthase|5-aminolevulinate synthase|5-aminolevulinic acid synthase 2|Alas 2|ALAS|ALAS E|erythroid|ALAS-E|ALASE|Aminolevulinate delta synthase 2|Aminolevulinic acid synthase 2|ANH1|ASB|Delta ALA synthase 2|Delta ALA synthetase|Delta aminolevulinate synthase 2|Delta aminolevulinate synthase|Delta-ALA synthase 2|Delta-aminolevulinate synthase 2|Erythroid specific ALAS|erythroid-specific|FLJ93603|HEM0_HUMAN|mitochondrial|OTTHUMP00000023388|OTTHUMP00000023389|OTTMUSP00000020679|RP23-338A17.1|SIDBA1|XLDPP|XLEPP|XLSA
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human 5-aminolevulinate synthase, erythroid-specific, mitochondrial protein (50-587AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ALAS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human