Anti-STARD13 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: STARD13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-663
- 77845-664
- Antigen Symbol:
- STARD13
- STARD13
- Antigen Synonyms:
- 46H23.2| ARHGAP37| Deleted in liver cancer 2 protein| Deleted in liver cancer protein 2| DLC-2| DLC2| GT650| Rho GTPase activating protein on chromosome 13q12| Rho GTPase-activating protein| STA13| STA13_HUMAN| StAR related lipid transfer (START) domain containing 13| StAR-related lipid transfer protein 13| StARD13| START domain-containing protein 13
- 46H23.2| ARHGAP37| Deleted in liver cancer 2 protein| Deleted in liver cancer protein 2| DLC-2| DLC2| GT650| Rho GTPase activating protein on chromosome 13q12| Rho GTPase-activating protein| STA13| STA13_HUMAN| StAR related lipid transfer (START) domain containing 13| StAR-related lipid transfer protein 13| StARD13| START domain-containing protein 13
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3M8
- Q9Y3M8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human StAR-related lipid transfer protein 13 protein (516-679AA)
- Recombinant Human StAR-related lipid transfer protein 13 protein (516-679AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice