Bacillus cereus Phospholipase C
Catalog # 80053-702
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- CAS No.:9001-86-9
- Species:Bacillus cereus
- Size:250 U
- Storage Conditions:Desiccated (hygroscopic), safe to freeze.
- Enzyme Name:Phospholipase C
- Enzyme Synonyms:PLC