Anti-CNR2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-343
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CNR2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P34972
- Antigen Synonyms:Cannabinoid receptor 2|Cannabinoid receptor 2 macrophage|CB 2|CB-2|CB2|CNR2_HUMAN|CNRII|CX 5|CX5|hCB2|testis-dominant CNR2 isoform CB2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cannabinoid receptor 2 protein (305-360AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human