SARS-CoV-2 Recombinant SARS-CoV-2 Spike Protein (from HEK293 Cells)
Catalog # 76834-310
Supplier: Antibodies.com
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Size:100 µg
- Storage Conditions:Upon delivery aliquot and store at −20...−70 °C. Avoid freeze/thaw cycles.
- Endotoxin Content:<0.1 ng/µg (1 IEU/µg), as measured by LAL method.
- Gene ID:UniprotID# P0DTC2
- Carrier-Free:N
- Protein Synonyms:SARS-CoV-2 Spike Protein
- Protein/Peptide Name:SARS-CoV-2 Spike Protein
- Purity:>95% (by SDS-PAGE)
- Molecular Weight:137 kDa (including tags)
- Sequence:VNLTTTTQLPPAYTNSFTRGVYYPDKVFRSSVLHSTQDLFLPFFSNVTWFHAIHVSGTNGTKRFDNPVLPFNDGVYFASTEKSNIIRGWIFGTTLDSKTQSLLIVNNATNVVIKVCEFQFCNDPFLGVYYHKNNKSWMKSEFRVYSSANNCTFEYVSQPFLMDLEGKQGNFKNLREFVFKNIDGYFKIYSKHTPINLVRDLPHGFSALEPLVDLPIGINITRFQTLLALHRSYLTPGDSSSGWTAGAAAYYVGYLQPRTFLLKYNENGTITDAVDCALDPLSETKCTLKSFTVEKGIYQTSNFRVQPTESIVRFPNITNLCPFGEVFNATRFASVYAWNRKRISNCVADYSVLYNSASFSTFKCYGVSPTKLNDLCFTNVYADSFVIRGDEVRQIAPGQTGKIADYNYKLPDDFTGCVIAWNSNNLDSKVGGNYNYRYRLFRKSNLKPFERDISTEIYQAGSTPCNGVQGFNCYFPLQSYGFQPTNGVGYQPYRVVVLSFELLHAPATVCGPKKSTNLVKNKCVNFNFNGLTGTGVLTESNKKFLPFQQFGRDIADTTDAVRDPQTLEILDITPCSFGGVSVITPGTNTSNQVAVLYQGVNCTEVPVAIHADQLTPTWRVYSTGSNVFQTRAGCLIGAEHVNNSYECDIPIGAGICASYQTQTNSRRGAGSVASQSIIAYTMSLGAENSVAYSNNSIAIPTNFTISVTTEILPVSMTKTSVDCTMYICGDSTECSNLLLQYGSFCTQLNRALTGIAVEQDKNTQEVFAQVKQIYKTPPIKDFGGFNFSQILPDPSKPSKRSPIEDLLFNKVTLADAGFIKQYGDCLGDIAARDLICAQKFNGLTVLPPLLTDEMIAQYTSALLAGTITSGWTFGAGPALQIPFPMQMAYRFNGIGVTQNVLYENQKLIANQFNSAIGKIQDSLSSTPSALGKLQDVVNQNAQALNTLVKQLSSNFGAISSVLNDILSRLDPPEAEVQIDRLITGRLQSLQTYVTQQLIRAAEIRASANLAATKMSECVLGQSKRVDFCGKGYHLMSFPQSAPHGVVFLHVTYVPAQEKNFTTAPAICHDGKAHFPREGVFVSNGTDWFVTQRNFYEPQIITTDNTFVSGNCDVVIGIVNNTVYDPLQPELDSFKEELDKYFKNHTSPDVDLGDISGINASVVNIQKEIDRLNEVAKNLNESLIDLQELGKYEQYIKGSGSGYIPEAPRDGQAYVRKDGEWVLLSTFLGSGHHHHHHHH
- Concentration:Lot specific
- Formulation:Supplied in phosphate buffered saline with 20% glycerol
- Shipping Temperature:Shipped on blue ice at 4 °C
- Tested Applications:WB, SDS-PAGE, Immunoassays, Functional Studies
Specifications
About this item
Partial length SARS-CoV-2 SARS-CoV-2 Spike protein, expressed in HEK293 cells, suitable for WB, SDS-PAGE, immunoassays, functional studies and is biologically active.
Caution: This product is for esearch use only. It is not for use in diagnostic or therapeutic procedures.