Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-NDUFS4 Rabbit Polyclonal Antibody
Anti-NDUFS4 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-NDUFS4 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77848-041
  • 77848-040
  • Antigen Symbol:
  • NDUFS4
  • NDUFS4
  • Antigen Synonyms:
  • AQDQ| CI 18| CI 18 kDa| CI AQDQ| CI-18 kDa| CI-AQDQ| Complex I 18 kDa| Complex I AQDQ| Complex I-18 kDa| Complex I-AQDQ| mitochondrial| mitochondrial respiratory chain complex I (18 KD subunit)| NADH coenzyme Q reductase| NADH dehydrogenase (ubiquinone) Fe S protein 4 18kDa| NADH dehydrogenase [ubiquinone] iron-sulfur protein 4| NADH dehydrogenase| NADH ubiquinone oxidoreductase 18 kDa subunit| NADH-ubiquinone oxidoreductase 18 kDa subunit| NDUFS4| NDUS4_HUMAN
  • AQDQ| CI 18| CI 18 kDa| CI AQDQ| CI-18 kDa| CI-AQDQ| Complex I 18 kDa| Complex I AQDQ| Complex I-18 kDa| Complex I-AQDQ| mitochondrial| mitochondrial respiratory chain complex I (18 KD subunit)| NADH coenzyme Q reductase| NADH dehydrogenase (ubiquinone) Fe S protein 4 18kDa| NADH dehydrogenase [ubiquinone] iron-sulfur protein 4| NADH dehydrogenase| NADH ubiquinone oxidoreductase 18 kDa subunit| NADH-ubiquinone oxidoreductase 18 kDa subunit| NDUFS4| NDUS4_HUMAN
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Mouse, Rat
  • Human, Mouse, Rat
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O43181
  • O43181
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Fusion protein of Human NDUFS4
  • Fusion protein of Human NDUFS4
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications