Anti-HOXA11/HOXD11 Rabbit Polyclonal Antibody
Catalog # 77847-268
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HOXA11/HOXD11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P31270
- Antigen Synonyms:Homeo box 1I|Homeo box A11|Homeobox A11|Homeobox protein Hox A11|Homeobox protein Hox-1I|Homeobox protein Hox-A11|Homeobox protein HOXA11|Hox 1I|HOX1|HOX1I|HOXA11|HXA11_HUMAN
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human HOXA11/D11.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000, IF: 1:100 to 1:500
Type:
Antigen: HOXA11/HOXD11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse