Anti-C1GALT1C1 Rabbit Recombinant Polyclonal Antibody
77852-041
New Product
- Antigen Symbol:ERCC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P18074
- Antigen Synonyms:TFIIH 80 kDa subunit|Basic transcription factor 2 80 kDa subunit|BTF2 p80|COFS 2|COFS2|CXPD|DNA excision repair protein ERCC 2|DNA excision repair protein ERCC-2|DNA repair protein complementing XP D cells|DNA repair protein complementing XP-D cells|EM9|ERCC 2|ERCC2|ERCC2_HUMAN|Excision repair 2|Excision repair cross complementing rodent repair deficiency complementation|Excision repair cross complementing rodent repair deficiency|complementation group 2|MAG|MGC102762|MGC126218|MGC126219|OTTHUMP00000045860|OTTHUMP00000045861|OTTHUMP00000045862|OTTHUMP00000045863|TFIIH basal transcription factor complex 80 kDa subunit|TFIIH Basal Transcription Factor Complex Helicase Subunit|TFIIH basal transcription factor complex helicase XPD subunit|TFIIH basal transcription factor complex p80 subunit|TFIIH p80|TTD|Xeroderma pigmentosum complementary group D|Xeroderma pigmentosum group D complementing protein|Xeroderma pigmentosum group D-complementing protein|XPD|XPDC
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:XPDC
- Purification:Antigen affinity purified
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human