Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Acetylcoenzyme A
Acetylcoenzyme A
Acetylcoenzyme A
Supplier:
MilliporeSigma
Certificates
Expand image
Acetylcoenzyme A
Supplier:
MilliporeSigma
Certificates
About this item
Specifications
CAS No.:
75520-41-1
Storage Conditions:
Safe to freeze, desiccated.
Enzyme Name:
Acetylcoenzyme A
Enzyme Synonyms:
Trilithium Salt
Purity:
≥83% (enzymatic)
Specifications
Related Information
Documentation
Certificates
Search by Lot Number
Documentation