Anti-TIMM8B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TIMM8B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-577
- 77846-578
- Antigen Symbol:
- TIMM8B
- TIMM8B
- Antigen Synonyms:
- DDP2|DDPL|TIM8B|Mitochondrial import inner membrane translocase subunit Tim8 B|DDP-like protein|Deafness dystonia protein 2
- DDP2|DDPL|TIM8B|Mitochondrial import inner membrane translocase subunit Tim8 B|DDP-like protein|Deafness dystonia protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5J9
- Q9Y5J9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Mitochondrial import inner membrane translocase subunit Tim8 B protein (33-83AA)
- Recombinant human Mitochondrial import inner membrane translocase subunit Tim8 B protein (33-83AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice