Anti-CCL7 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: CCL7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-710
- 77843-709
- Antigen Symbol:
- CCL7
- CCL7
- Antigen Synonyms:
- C-C motif chemokine 7|CCL7_HUMAN|Chemokine CC motif ligand 7|FIC|MARC|MCP-3|Monocyte chemoattractant protein 3|Monocyte chemotactic protein 3|NC28|RP23-350G1.4|SCYA6|SCYA7|Small-inducible cytokine A7
- C-C motif chemokine 7|CCL7_HUMAN|Chemokine CC motif ligand 7|FIC|MARC|MCP-3|Monocyte chemoattractant protein 3|Monocyte chemotactic protein 3|NC28|RP23-350G1.4|SCYA6|SCYA7|Small-inducible cytokine A7
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P80098
- P80098
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human CCL7
- Synthetic peptide of Human CCL7
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice