Anti-add Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77850-723
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:add
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:E. coli
- Size:50 µg
- Form:Liquid
- Gene ID:P22333
- Antigen Synonyms:b1623|JW1615Adenosine deaminase|EC 3.5.4.4|Adenosine aminohydrolase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Escherichia coli Adenosine deaminase protein (1-333AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: add
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli