Anti-CHN2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-086
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CHN2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P52757
- Antigen Synonyms:ARHGAP3|BCH|Beta-chimaerin|Beta-chimerin|Rho GTPase-activating protein 3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Beta-chimaerin protein (1-91AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:500 to 1:1000
Type:
Antigen: CHN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human