Anti-ARFIP2 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
77851-337
New Product
- Antigen Symbol:ARFIP2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P53365
- Antigen Synonyms:ADP ribosylation factor interacting protein 2|ADP-ribosylation factor-interacting protein 2|Arfaptin 2|Arfaptin-2|ARFP2_HUMAN|FLJ18046|FLJ18697|FLJ99239|Partner of RAC1|POR1|Protein POR1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Arfaptin-2 protein (20-295AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
0
Type:
Antigen: ARFIP2
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human