Anti-HRH4 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HRH4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-122
- 77843-121
- Antigen Symbol:
- HRH4
- HRH4
- Antigen Synonyms:
- AXOR35|BG26|G protein coupled receptor 105|G-protein coupled receptor 105|GPCR105|GPRv53|H4|H4R|HH4R|Histamine H4 receptor|Histamine receptor H4|Hrh4|HRH4_HUMAN|Pfi-013|SP9144
- AXOR35|BG26|G protein coupled receptor 105|G-protein coupled receptor 105|GPCR105|GPRv53|H4|H4R|HH4R|Histamine H4 receptor|Histamine receptor H4|Hrh4|HRH4_HUMAN|Pfi-013|SP9144
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H3N8
- Q9H3N8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Histamine H4 receptor protein (204-292AA)
- Recombinant Human Histamine H4 receptor protein (204-292AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice