Anti-BLK Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-898
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:BLK
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P51451
- Antigen Synonyms:B lymphocyte kinase|B lymphoid tyrosine kinase|BLK nonreceptor tyrosine kinase|BLK_HUMAN|EC 2.7.10.2|MGC 10442|MGC10442|MODY 11|MODY11|P55 p55-Tyrosine kinase B lymphocyte specific|Tyrosine protein kinase|Tyrosine-protein kinase BLK
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Tyrosine-protein kinase Blk protein (1-280AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: BLK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse