Anti-MDH2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77859-608
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MDH2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P40926
- Antigen Synonyms:M MDH|Malate dehydrogenase 2|NAD (mitochondrial)|Malate dehydrogenase|mitochondrial|MDH|MDHM_HUMAN|MGC:3559|Mitochondrial malate dehydrogenase 2|NAD|Mor 1|MOR1
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Malate dehydrogenase, mitochondrial protein (59-338AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: MDH2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse