Anti-CCL27 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: CCL27
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-692
- 77845-693
- Antigen Symbol:
- CCL27
- CCL27
- Antigen Synonyms:
- ALP| C-C motif chemokine 27| CC chemokine ILC| Ccl27| CCL27_HUMAN| Chemokine (C-C motif) ligand 27| CTACK| CTAK| Cutaneous T cell attracting chemokine| Cutaneous T-cell-attracting chemokine| ESkine| IL 11 R alpha locus chemokine| IL-11 R-alpha-locus chemokine| ILC| PESKY| SCYA27| Skinkine| Small inducible cytokine A27 | small inducible cytokine subfamily A (Cys-Cys)| member 27| Small-inducible cytokine A27
- ALP| C-C motif chemokine 27| CC chemokine ILC| Ccl27| CCL27_HUMAN| Chemokine (C-C motif) ligand 27| CTACK| CTAK| Cutaneous T cell attracting chemokine| Cutaneous T-cell-attracting chemokine| ESkine| IL 11 R alpha locus chemokine| IL-11 R-alpha-locus chemokine| ILC| PESKY| SCYA27| Skinkine| Small inducible cytokine A27 | small inducible cytokine subfamily A (Cys-Cys)| member 27| Small-inducible cytokine A27
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y4X3
- Q9Y4X3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human C-C motif chemokine 27 protein (25-112AA)
- Recombinant Human C-C motif chemokine 27 protein (25-112AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice