Anti-ANXA13 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-169
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ANXA13
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Dog
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q29471
- Antigen Synonyms:ANX13Annexin A13|Annexin XIII|Annexin-13|Intestine-specific annexin|ISA
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Dog Annexin A13 protein (2-316AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: ANXA13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog