Anti-PLAGL1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PLAGL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-985
- 77844-984
- Antigen Symbol:
- PLAGL1
- PLAGL1
- Antigen Synonyms:
- DKFZp781P1017|Lost on transformation 1|Lost on Transformation|LOT-1|LOT1|MGC126275|MGC126276|PLAG like 1|PLAL1_HUMAN|Pleiomorphic adenoma gene like 1|Pleiomorphic adenoma gene like protein 1|Pleiomorphic adenoma-like protein 1|Tumor supressor ZAC|ZAC|ZAC tumor supressor|ZAC1|Zinc finger protein PLAGL1
- DKFZp781P1017|Lost on transformation 1|Lost on Transformation|LOT-1|LOT1|MGC126275|MGC126276|PLAG like 1|PLAL1_HUMAN|Pleiomorphic adenoma gene like 1|Pleiomorphic adenoma gene like protein 1|Pleiomorphic adenoma-like protein 1|Tumor supressor ZAC|ZAC|ZAC tumor supressor|ZAC1|Zinc finger protein PLAGL1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UM63
- Q9UM63
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Zinc finger protein PLAGL1 protein (237-372AA)
- Recombinant Human Zinc finger protein PLAGL1 protein (237-372AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together