Anti-IPO13 Rabbit Recombinant Polyclonal Antibody
77858-163
New Product
- Antigen Symbol:IPO13
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O94829
- Antigen Synonyms:Imp 13|Imp13|Importin-13|Importin13|IPO 13|IPO13_HUMAN|Kap 13|Kap13|Karyopherin 13|Karyopherin-13|Karyopherin13|KIAA0274|Late gestation lung 2|Late gestation lung 2 protein|Lgl2|Ran binding protein 13|Ran-binding protein 13|RanBP 13|RanBP13
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Importin-13 protein (501-700AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: IPO13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human