Anti-AP2M1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-829
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AP2M1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q96CW1
- Antigen Synonyms:Adapter-related protein complex 2 mu subunit|Adaptin mu 1|Adaptin-mu2|Adaptor protein complex AP 2 subunit mu|Adaptor protein complex AP-2 subunit mu|Adaptor related protein complex 2 mu 1 subunit|AP 2 mu 2 chain|AP-2 complex subunit mu|AP-2 mu chain|Ap2m1|AP2M1_HUMAN|AP50|CLAPM1|Clathrin adaptor complex AP2 mu subunit|Clathrin assembly protein complex 2 medium chain|Clathrin associated/assembly/adaptor protein medium 1|Clathrin coat adaptor protein AP50|Clathrin coat assembly protein AP50|Clathrin coat associated protein AP50|Clathrin coat-associated protein AP50|HA2 50 kDa subunit|mu2|Plasma membrane adaptor AP-2 50 kDa protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human AP-2 complex subunit mu protein (1-435AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: AP2M1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human