Anti-CYBRD1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-853
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CYBRD1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q53TN4
- Antigen Synonyms:CYBR 1|CYBR1|CYBR1_HUMAN|CYBRD 1|Cytochrome b reductase 1|DCYTB|DKFZp564E227|Duodenal cytochrome b|Ferric chelate reductase 3|Ferric-chelate reductase 3|FLJ23462|FRRS3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cytochrome b reductase 1 protein (220-286AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IF:1:200 to 1:500
Type:
Antigen: CYBRD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human